This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01257340_g1
See other SV2A GE Assays ›
Gene Symbol:
Sv2a
Gene Name:
synaptic vesicle glycoprotein 2 a
Gene Aliases:
AI746429, mKIAA0736
Chromosome Location:
Chr.3: 96181227 - 96195183 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01257340_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sv2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022030.3 NP_071313.1 8 - 9 1797 66
GenBank mRNA AK028318.1 - 8 - 9 1742 66
AK080502.1 - 8 - 9 1750 66
AK080525.1 - 8 - 9 1839 66
AK122360.1 - 8 - 9 1833 66
AK144166.1 - 8 - 9 1820 66
BC026494.1 - 2 - 3 144 66
BC046587.1 - 8 - 9 1797 6493509 66
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK158936.1 - Chr. - - [-] on Build 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000928.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sv2a

Gene Ontology Categories:

Function(s) Process(es)

transport
neurotransmitter transport
cellular calcium ion homeostasis
chemical synaptic transmission
transmembrane transport
receptor activity
transporter activity
protein kinase binding
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300007435.mm.1 NM_022030.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300007435.mm.1 NM_022030.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300007435.mm.1 TC0300000928.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0