This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01257361_g1
See other ATP5A1 GE Assays ›
Gene Symbol:
Atp5a1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1
Gene Aliases:
AI035633, AL022851, AL023067, Atpm, D18Ertd206e, Mom2
Chromosome Location:
Chr.18: 77773768 - 77782869 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01257361_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007505.2 NP_031531.1 11 - 12 1858 64
GenBank mRNA AK043976.1 - 11 - 12 1626 64
AK076572.1 - 11 - 12 1626 64
AK098155.1 - 11 - 12 1636 64
AK146797.1 - 11 - 12 1524 64
AK150426.1 - 11 - 12 1629 64
AK150843.1 - 11 - 12 1626 64
AK151004.1 - 11 - 12 1626 64
AK151128.1 - 11 - 12 1635 64
AK151224.1 - 11 - 12 1633 64
AK151920.1 - 11 - 12 1630 64
AK152054.1 - 11 - 12 1637 64
AK152890.1 - 11 - 12 1629 64
AK153167.1 - 11 - 12 1630 64
AK153282.1 - 11 - 12 1626 64
AK159491.1 - 11 - 12 1626 64
AK159540.1 - 11 - 12 1626 64
AK159758.1 - 11 - 12 1629 64
AK160043.1 - 11 - 12 1636 64
AK164110.1 - 11 - 12 1629 64
AK164193.1 - 9 - 10 1332 64
AK166709.1 - 11 - 12 1629 64
AK166812.1 - 11 - 12 1642 64
AK167159.1 - 11 - 12 1626 64
AK167863.1 - 11 - 12 1606 64
AK168198.1 - 11 - 12 1636 64
AK168369.1 - 11 - 12 1628 64
AK168617.1 - 11 - 12 1626 64
AK168879.1 - 11 - 12 1626 64
AK168890.1 - 11 - 12 1626 64
AK168932.1 - 11 - 12 1626 64
AK169080.1 - 11 - 12 1629 64
AK169084.1 - 11 - 12 1629 64
AK169105.1 - 11 - 12 1626 64
AK169142.1 - 11 - 12 1671 64
AK169300.1 - 11 - 12 1626 64
AK169308.1 - 11 - 12 1631 64
AK169414.1 - 11 - 12 1629 64
AK180896.1 - 1 - 2 34 64
AK214653.1 - 1 - 2 34 64
AK214982.1 - 1 - 2 34 64
AK215077.1 - 1 - 2 34 64
BC014854.1 - 11 - 12 1613 4500893 64
BC020417.1 - 11 - 12 1613 64
BC055892.1 - 11 - 12 2063 64
L01062.1 - 11 - 12 1613 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000833.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5a1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endothelial cell proliferation
lipid metabolic process
ATP biosynthetic process
transport
ion transport
embryo development
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
proton transport
ATP metabolic process
hydrogen ion transmembrane transport
nucleotide binding
ATP binding
hydrolase activity, acting on acid anhydrides, catalyzing transmembrane movement of substances
ATPase activity
MHC class I protein binding
ADP binding
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31849680 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31849682 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31849681 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041340 BY465601.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041340 AK169308.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041340 AK169084.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041340 AK168932.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2041340 AK168369.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437160 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408407 AK041275.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6144 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306196 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214079 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118807 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160492 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041340 BY465601.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041340 AK169308.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041340 AK169084.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041340 AK168932.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2041340 AK168369.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402496 L01062.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P674489 L01062.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P674489 BC020417.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P674489 BC014854.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P674489 AK214653.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P674489 AK214982.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849690 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849699 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849693 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849687 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849683 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849697 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849684 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849686 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849695 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849689 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849682 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849691 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849680 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31849688 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160492 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09060215 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09060234 - -
Whole Mouse Genome G4122A A_52_P674489 L01062.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P674489 BC020417.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P674489 BC014854.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P674489 AK214653.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P674489 AK214982.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P402496 L01062.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133545 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140213 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09060234 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160492 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13845993 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13837984 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31849680 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31849681 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31849682 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849678 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849739 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849712 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849719 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849774 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849742 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849680 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849753 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849635 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849738 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849705 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849747 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849730 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849710 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849733 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849728 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849734 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849736 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849687 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849729 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849691 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849726 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849745 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849684 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849689 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849679 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849723 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849695 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849707 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849715 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849704 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31849767 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849697 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849684 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849683 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849678 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849688 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849707 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849738 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849703 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849704 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849687 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849699 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849693 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849676 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849679 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849690 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849709 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849741 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849714 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849742 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849721 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849686 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14069462 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849675 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849715 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849695 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849669 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849717 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849743 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849728 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13630026 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849660 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849710 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849689 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849701 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849671 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849674 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849673 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849705 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849719 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849718 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849739 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849691 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849665 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13890350 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849670 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31849723 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14069461 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10456903 - -
Affymetrix Mouse Gene 1.0 ST Array 10456898 - -
Affymetrix Mouse Gene 1.0 ST Array 10456896 - -
Affymetrix Mouse Gene 1.0 ST Array 10456893 - -
Affymetrix Mouse Gene 1.0 ST Array 10456895 - -
Affymetrix Mouse Gene 1.0 ST Array 10456894 - -
Affymetrix Mouse Gene 1.0 ST Array 10456900 - -
Affymetrix Mouse Gene 1.0 ST Array 10456897 - -
Affymetrix Mouse Gene 1.0 ST Array 10456892 - -
Affymetrix Mouse Gene 1.0 ST Array 10456901 - -
Affymetrix Mouse Gene 1.0 ST Array 10456899 - -
Affymetrix Mouse Gene 1.0 ST Array 10456902 - -
Affymetrix HT MG-430 PM Array Plate 1449710_PM_s_at - -
Affymetrix HT MG-430 PM Array Plate 1420037_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1423111_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10456891 - -
Mouse Genome 430 2.0 Array 1449710_PM_s_at - -
Mouse Genome 430 2.0 Array 1449710_s_at - -
Mouse Genome 430 2.0 Array 1420037_at - -
Mouse Genome 430 2.0 Array 1423111_at - -
Mouse Genome 430 2.0 Array 1420037_PM_at - -
Mouse Genome 430 2.0 Array 1423111_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11946_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 11946_PM_at - -
Affymetrix Mouse Clariom S Array PSR1800006094.mm.1 NM_007505.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006094.mm.1 NM_007505.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800006094.mm.1 TC1800000833.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800006096.mm.1 NM_007505.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006096.mm.1 NM_007505.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800006096.mm.1 TC1800000833.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800006095.mm.1 NM_007505.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006095.mm.1 NM_007505.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800006095.mm.1 TC1800000833.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800006097.mm.1 NM_007505.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006097.mm.1 NM_007505.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800006097.mm.1 TC1800000833.mm.1 MTA TC
Mouse Expression Set 430 1423111_at - -
Mouse Expression Set 430 1423111_PM_at - -
Murine Genome U74v2 Set 100753_at - -
Murine Genome U74v2 Set 100753_PM_at - -
Mouse Genome 430A 2.0 Array 1423111_at - -
Mouse Genome 430A 2.0 Array 1423111_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1423111_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1423111_at - -
Affymetrix HT Mouse Genome 430A Array 1449710_s_at - -
Affymetrix HT Mouse Genome 430A Array 1449710_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1420037_at - -
Affymetrix HT Mouse Genome 430A Array 1420037_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 1990722 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001990722 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2633229 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2633229 - -
mouse-6 v1.1 (Illumina) scl51737.13.1_28-S - -
Illumina Sentrix mouse-6.v1 bead array 1990722 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl51737.13.1_28-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990722 - -
MouseRef-8 v2 Expression BeadChip ILMN_2633229 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2633229 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl51737.13.1_28-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0