This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01257365_g1
See other ATP5A1 GE Assays ›
Gene Symbol:
Atp5a1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1
Gene Aliases:
AI035633, AL022851, AL023067, Atpm, D18Ertd206e, Mom2
Chromosome Location:
Chr.18: 77773768 - 77782869 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01257365_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007505.2 NP_031531.1 5 - 6 935 76
GenBank mRNA AK043976.1 - 5 - 6 703 76
AK076572.1 - 5 - 6 703 76
AK098155.1 - 5 - 6 713 76
AK146797.1 - 5 - 6 706 76
AK150426.1 - 5 - 6 706 76
AK150843.1 - 5 - 6 703 76
AK151128.1 - 5 - 6 712 76
AK151920.1 - 5 - 6 707 76
AK152054.1 - 5 - 6 714 76
AK152890.1 - 5 - 6 706 76
AK153167.1 - 5 - 6 707 76
AK153282.1 - 5 - 6 703 76
AK157529.1 - 3 - 4 484 76
AK159491.1 - 5 - 6 703 76
AK159540.1 - 5 - 6 703 76
AK159758.1 - 5 - 6 706 76
AK160043.1 - 5 - 6 713 76
AK164110.1 - 5 - 6 706 76
AK164193.1 - 3 - 4 409 76
AK166709.1 - 5 - 6 706 76
AK166812.1 - 5 - 6 719 76
AK167159.1 - 5 - 6 703 76
AK167863.1 - 5 - 6 683 76
AK168198.1 - 5 - 6 713 76
AK168617.1 - 5 - 6 703 76
AK168879.1 - 5 - 6 703 76
AK168890.1 - 5 - 6 703 76
AK168932.1 - 5 - 6 703 76
AK169080.1 - 5 - 6 706 76
AK169084.1 - 5 - 6 706 76
AK169105.1 - 5 - 6 703 76
AK169142.1 - 5 - 6 748 76
AK169300.1 - 5 - 6 703 76
AK169308.1 - 5 - 6 708 76
AK169414.1 - 5 - 6 706 76
BC014854.1 - 5 - 6 690 4500893 76
BC055892.1 - 5 - 6 1140 76
L01062.1 - 5 - 6 690 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000833.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5a1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endothelial cell proliferation
lipid metabolic process
ATP biosynthetic process
transport
ion transport
embryo development
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
proton transport
ATP metabolic process
hydrogen ion transmembrane transport
nucleotide binding
ATP binding
hydrolase activity, acting on acid anhydrides, catalyzing transmembrane movement of substances
ATPase activity
MHC class I protein binding
ADP binding
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800006085.mm.1 NM_007505.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800006085.mm.1 NM_007505.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800006085.mm.1 TC1800000833.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0