This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01257987_m1
See other SCP2 GE Assays ›
Gene Symbol:
Scp2
Gene Name:
sterol carrier protein 2, liver
Gene Aliases:
AA409774, AA409893, C76618, C79031, NSL-TP, SCP-2, SCP-X, SCPX, ns-LTP
Chromosome Location:
Chr.4: 108043830 - 108144971 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01257987_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Scp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011327.4 NP_035457.1 1 - 2 173 99
GenBank mRNA AK004860.2 - 1 - 2 103 99
BC018384.1 - 1 - 2 128 3710340 99
BC034613.1 - 1 - 2 94 5123611 99
DN176201.1 - 1 - 2 173 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003234.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Scp2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

acyl-CoA metabolic process
steroid biosynthetic process
progesterone biosynthetic process
transport
lipid transport
peroxisome organization
aging
metabolic process
positive regulation of steroid biosynthetic process
phospholipid transport
cholesterol transport
positive regulation of intracellular cholesterol transport
inositol trisphosphate biosynthetic process
positive regulation of apoptotic process
positive regulation of cholesterol biosynthetic process
positive regulation of steroid metabolic process
protein localization to plasma membrane
lipid hydroperoxide transport
positive regulation of cholesterol import
fatty-acyl-CoA binding
catalytic activity
acetyl-CoA C-acyltransferase activity
receptor binding
protein binding
lipid binding
phosphatidylcholine transporter activity
phosphatidylinositol transporter activity
cholesterol binding
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
cholesterol transporter activity
propanoyl-CoA C-acyltransferase activity
long-chain fatty acyl-CoA binding
protein homodimerization activity
protein heterodimerization activity
propionyl-CoA C2-trimethyltridecanoyltransferase activity
oleic acid binding
phosphatidylethanolamine transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400025751.mm.1 TC0400003234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400025751.mm.1 NM_011327.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400025751.mm.1 NM_011327.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400025746.mm.1 TC0400003234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400025746.mm.1 NM_011327.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400025746.mm.1 NM_011327.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400025743.mm.1 TC0400003234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400025743.mm.1 NM_011327.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400025743.mm.1 NM_011327.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0