This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01258325_m1
See other FGF1 GE Assays ›
Gene Symbol:
Fgf1
Gene Name:
fibroblast growth factor 1
Gene Aliases:
Dffrx, Fam, Fgf-1, Fgfa
Chromosome Location:
Chr.18: 38838672 - 38929470 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01258325_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010197.3 NP_034327.1 1 - 2 149 80
GenBank mRNA AK035330.1 - 1 - 2 150 80
U67610.1 - 1 - 2 115 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK035330.1, NM_010197.3, U67610.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fgf1

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
organ induction
positive regulation of protein phosphorylation
multicellular organism development
cell proliferation
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
cell differentiation
lung development
positive regulation of cell migration
cellular response to heat
positive regulation of MAP kinase activity
positive regulation of cholesterol biosynthetic process
positive regulation of angiogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of epithelial cell proliferation
positive regulation of cell division
cardiac muscle cell proliferation
branch elongation involved in ureteric bud branching
vasculogenesis involved in coronary vascular morphogenesis
mesonephric epithelium development
positive regulation of intracellular signal transduction
regulation of endothelial cell chemotaxis to fibroblast growth factor
receptor binding
fibroblast growth factor receptor binding
protein binding
growth factor activity
heparin binding
Hsp70 protein binding
S100 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800009196.mm.1 NM_010197.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800009196.mm.1 NM_010197.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800009196.mm.1 TC1800001237.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800009191.mm.1 NM_010197.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800009191.mm.1 NM_010197.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800009191.mm.1 TC1800001237.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-4kcjf:80/100.66.78.36:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline