This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01258503_m1
See other AFAP1L1 GE Assays ›
Gene Symbol:
Afap1l1
Gene Name:
actin filament associated protein 1-like 1
Gene Aliases:
AI173486
Chromosome Location:
Chr.18: 61730262 - 61786662 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01258503_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Afap1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178928.4 NP_849259.2 9 - 10 1089 73
GenBank mRNA AK035227.1 - 9 - 10 1089 73
AK053714.1 - 9 - 10 1088 73
BC094505.1 - 6 - 7 527 73
BC138923.1 - 9 - 10 1050 8861945 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001430.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Afap1l1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800010502.mm.1 NM_178928.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010502.mm.1 NM_178928.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800010502.mm.1 TC1800001430.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800010504.mm.1 NM_178928.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010504.mm.1 NM_178928.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800010504.mm.1 TC1800001430.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0