This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01258787_m1
See other ABLIM3 GE Assays ›
Gene Symbol:
Ablim3
Gene Name:
actin binding LIM protein family, member 3
Gene Aliases:
AW990719, D930036B08Rik
Chromosome Location:
Chr.18: 61799392 - 61911857 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01258787_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ablim3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164491.1 NP_001157963.1 14 - 15 1562 88
NM_198649.3 NP_941051.2 13 - 14 1881 88
GenBank mRNA AK086538.1 - 13 - 14 1662 88
AK173040.1 - 13 - 14 1439 88
BC060275.1 - 14 - 15 1533 6822626 88
BC094229.1 - 13 - 14 1855 30534236 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001431.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ablim3

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
cytoskeleton organization
lamellipodium assembly
actin cytoskeleton organization
cilium assembly
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein targeting to mitochondrion
molecular_function
actin binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800010532.mm.1 NM_001164491.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010532.mm.1 NM_001164491.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010532.mm.1 NM_198649.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800010532.mm.1 NM_198649.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800010532.mm.1 TC1800001431.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0