This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01258860_g1
See other TNFRSF18 GE Assays ›
Gene Symbol:
Tnfrsf18
Gene Name:
tumor necrosis factor receptor superfamily, member 18
Gene Aliases:
AITR, Gitr
Chromosome Location:
Chr.4: 156026342 - 156028891 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01258860_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009400.2 NP_033426.1 2 - 3 315 65
NM_021985.2 NP_068820.1 2 - 3 315 65
GenBank mRNA AF229432.1 - 2 - 3 315 65
AF229433.1 - 2 - 3 315 65
AF229434.1 - 2 - 3 315 65
AK020762.1 - 2 - 3 336 65
AK081878.1 - 2 - 3 304 65
AY157832.1 - 2 - 3 315 65
AY157833.1 - 2 - 3 315 65
U82534.1 - 2 - 3 315 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002104.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf18

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of leukocyte migration
apoptotic process
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of cell adhesion
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400017741.mm.1 NM_009400.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017741.mm.1 NM_009400.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017741.mm.1 NM_021985.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017741.mm.1 NM_021985.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400017741.mm.1 TC0400002104.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0