This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01258910_g1
See other ALAD GE Assays ›
Gene Symbol:
Alad
Gene Name:
aminolevulinate, delta-, dehydratase
Gene Aliases:
ALADH, Lv
Chromosome Location:
Chr.4: 62505984 - 62520063 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01258910_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Alad
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276446.1 NP_001263375.1 9 - 10 902 90
NM_008525.4 NP_032551.3 9 - 10 991 90
GenBank mRNA AK002300.1 - 9 - 10 887 90
AK032908.1 - 9 - 10 847 90
AK167673.1 - 9 - 10 822 90
AK168119.1 - 9 - 10 858 90
AK168132.1 - 9 - 10 863 90
BC018236.1 - 8 - 9 1409 3992605 90
BC055930.1 - 9 - 10 991 4207420 90
U89407.1 - 6 - 7 483 90
X13752.1 - 8 - 9 719 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002791.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Alad

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

porphyrin-containing compound biosynthetic process
heme biosynthetic process
response to nutrient
metabolic process
response to organic substance
response to metal ion
response to zinc ion
tetrapyrrole biosynthetic process
response to drug
response to ethanol
response to arsenic-containing substance
response to cadmium ion
protein homooligomerization
cellular response to interleukin-4
catalytic activity
porphobilinogen synthase activity
zinc ion binding
lyase activity
lead ion binding
identical protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400022914.mm.1 TC0400002791.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400022914.mm.1 NM_001276446.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022914.mm.1 NM_008525.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400022913.mm.1 TC0400002791.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400022913.mm.1 NM_001276446.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400022913.mm.1 NM_008525.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0