This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01259104_m1
See other GLB1L GE Assays ›
Gene Symbol:
Glb1l
Gene Name:
galactosidase, beta 1-like
Gene Aliases:
4833408P15Rik
Chromosome Location:
Chr.1: 75198235 - 75210816 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01259104_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glb1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029010.1 NP_083286.1 1 - 2 184 55
GenBank mRNA AK029925.1 - 1 - 2 184 55

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK029925.1, NM_029010.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

galactosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
metabolic process
molecular_function
hydrolase activity, hydrolyzing O-glycosyl compounds
beta-galactosidase activity
lytic transglycosylase activity
mannosidase activity
mannosyl-oligosaccharide mannosidase activity
galactosidase activity
glucosidase activity
trehalase activity
fucosidase activity
hexosaminidase activity
amylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
dextrin alpha-glucosidase activity
starch alpha-glucosidase activity
beta-glucanase activity
beta-6-sulfate-N-acetylglucosaminidase activity
glucan endo-1,4-beta-glucosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100021187.mm.1 NM_029010.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100021187.mm.1 NM_029010.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100021187.mm.1 TC0100002603.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100021192.mm.1 NM_029010.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100021192.mm.1 NM_029010.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100021192.mm.1 TC0100002603.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100021193.mm.1 NM_029010.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100021193.mm.1 NM_029010.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100021193.mm.1 TC0100002603.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-cns84:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline