This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01259143_g1
See other BCS1L GE Assays ›
Gene Symbol:
Bcs1l
Gene Name:
BCS1-like (yeast)
Gene Aliases:
9130022O19Rik
Chromosome Location:
Chr.1: 74588294 - 74592443 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01259143_g1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bcs1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK012324.1 - 2 - 3 514 60
AK078925.1 - 2 - 3 526 60
AK079385.1 - 2 - 3 522 60
BC019781.1 - 2 - 3 414 4036047 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000638.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20349430 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096485 AK212348.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096485 AK079385.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096485 NM_025784.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096486 BC019781.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096486 AK212348.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096486 AK012324.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096486 AK078925.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2096486 AK079385.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124972 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569974 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096485 AK212348.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096485 AK079385.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096485 NM_025784.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096486 BC019781.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096486 AK212348.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096486 AK012324.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096486 AK078925.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2096486 AK079385.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P104782 BC019781.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P104782 AK212348.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P104782 AK012324.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P104782 AK078925.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P104782 AK079385.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349430 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349410 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349428 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349431 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349443 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349446 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349449 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349422 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349465 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349451 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349455 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349401 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349417 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349440 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349453 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349409 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349415 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349424 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349425 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349398 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349438 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349447 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349413 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349426 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349444 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349467 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349436 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349420 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349419 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349432 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349406 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349460 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349461 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349463 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349403 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349405 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349458 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349400 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20349412 - -
Whole Mouse Genome G4122A A_52_P104782 BC019781.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P104782 AK212348.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P104782 AK012324.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P104782 AK078925.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P104782 AK079385.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101017 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100961 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00592535 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00776598 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00776600 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20349430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349507 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349451 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349453 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349488 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349501 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349493 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349460 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349458 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349489 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349495 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349446 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349428 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349465 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349525 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20349531 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349436 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349447 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349446 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00592535 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349443 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00592541 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349463 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00592543 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349451 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349438 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349460 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00776598 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349458 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349467 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00592546 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349440 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00592528 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349430 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00463538 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00463541 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P20349432 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P00463535 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10347426 - -
Affymetrix Mouse Gene 1.0 ST Array 10347421 - -
Affymetrix Mouse Gene 1.0 ST Array 10347418 - -
Affymetrix Mouse Gene 1.0 ST Array 10347424 - -
Affymetrix Mouse Gene 1.0 ST Array 10347419 - -
Affymetrix Mouse Gene 1.0 ST Array 10347425 - -
Affymetrix Mouse Gene 1.0 ST Array 10347422 - -
Affymetrix Mouse Gene 1.0 ST Array 10347420 - -
Affymetrix Mouse Gene 1.0 ST Array 10347423 - -
Affymetrix HT MG-430 PM Array Plate 1451541_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10347417 - -
Mouse Genome 430 2.0 Array 1451541_at - -
Mouse Genome 430 2.0 Array 1451541_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66821_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66821_PM_at - -
Affymetrix Mouse Clariom S Array PSR0100005220.mm.1 NM_025784.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100005220.mm.1 TC0100000638.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100005221.mm.1 NM_025784.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100005221.mm.1 TC0100000638.mm.1 MTA TC
Mouse Expression Set 430 1451541_at - -
Mouse Expression Set 430 1451541_PM_at - -
Mouse Genome 430A 2.0 Array 1451541_at - -
Mouse Genome 430A 2.0 Array 1451541_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1451541_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1451541_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 5360102 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005360102 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248953 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1248953 - -
mouse-6 v1.1 (Illumina) scl17795.8.1_1-S - -
Illumina Sentrix mouse-6.v1 bead array 5360102 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl17795.8.1_1-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360102 - -
MouseRef-8 v2 Expression BeadChip ILMN_2691345 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2691345 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl17795.8.1_1-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0