This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01259194_m1
See other BCAT1 GE Assays ›
Gene Symbol:
Bcat1
Gene Name:
branched chain aminotransferase 1, cytosolic
Gene Aliases:
BCATc, Eca39
Chromosome Location:
Chr.6: 144993835 - 145076157 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01259194_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bcat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007532.5 NP_031558.3 1 - 2 265 89
GenBank mRNA AK166971.1 - 1 - 2 201 89
AK167232.1 - 1 - 2 192 89
AK167368.1 - 1 - 2 111 89
AK168249.1 - 1 - 2 295 89
BC053706.1 - 1 - 2 197 30063465 89
BY781109.1 - 1 - 2 89
X17502.1 - 1 - 2 189 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK166971.1, AK167232.1, AK167368.1, AK168249.1, BC053706.1, NM_007532.5, X17502.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bcat1

Panther Classification:

Molecular Function -

transaminase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
cellular amino acid biosynthetic process
branched-chain amino acid metabolic process
branched-chain amino acid biosynthetic process
branched-chain amino acid catabolic process
mitotic G1 DNA damage checkpoint
cellular response to UV
catalytic activity
branched-chain-amino-acid transaminase activity
transaminase activity
transferase activity
identical protein binding
L-leucine transaminase activity
L-valine transaminase activity
L-isoleucine transaminase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600026882.mm.1 TC0600003412.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600026882.mm.1 NM_007532.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600026882.mm.1 NM_007532.5 Refseq
Affymetrix Mouse Clariom S Array PSR0600026877.mm.1 TC0600003412.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600026877.mm.1 NM_001024468.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600026877.mm.1 NM_001024468.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600026877.mm.1 NM_007532.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600026877.mm.1 NM_007532.5 Refseq
Affymetrix Mouse Clariom S Array PSR0600026881.mm.1 TC0600003412.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600026881.mm.1 NM_007532.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600026881.mm.1 NM_007532.5 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 526689 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0