This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01261113_m1
See other GPAM GE Assays ›
Gene Symbol:
Gpam
Gene Name:
glycerol-3-phosphate acyltransferase, mitochondrial
Gene Aliases:
GPAT, GPAT1
Chromosome Location:
Chr.19: 55067635 - 55127233 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01261113_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008149.3 NP_032175.2 8 - 9 945 85
GenBank mRNA AK047295.1 - 8 - 9 946 85
AK137067.1 - 10 - 11 1226 85
AK173202.1 - 8 - 9 970 85
BC019201.1 - 8 - 9 950 5041120 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001691.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpam

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
acyl-CoA metabolic process
triglyceride metabolic process
glycerophospholipid metabolic process
metabolic process
phospholipid biosynthetic process
response to glucose
positive regulation of triglyceride biosynthetic process
response to activity
triglyceride biosynthetic process
cellular response to insulin stimulus
positive regulation of multicellular organism growth
positive regulation of activated T cell proliferation
cellular lipid metabolic process
regulation of cytokine secretion
defense response to virus
fatty acid homeostasis
phospholipid homeostasis
negative regulation of activation-induced cell death of T cells
interleukin-2 secretion
glycerol-3-phosphate O-acyltransferase activity
O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900014837.mm.1 NM_008149.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900014837.mm.1 NM_008149.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900014837.mm.1 TC1900001691.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0