This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01261327_m1
See other MGST1 GE Assays ›
Gene Symbol:
Mgst1
Gene Name:
microsomal glutathione S-transferase 1
Gene Aliases:
1500002K10Rik, Gst
Chromosome Location:
Chr.6: 138140537 - 138156755 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01261327_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mgst1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019946.4 NP_064330.2 2 - 3 225 60
GenBank mRNA AF159050.1 - 1 - 2 124 60
AK002800.1 - 2 - 3 226 60
AK005122.1 - 2 - 3 226 60
BC009155.1 - 1 - 2 164 2655738 60

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mgst1

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
response to organonitrogen compound
response to lipopolysaccharide
response to drug
oxidation-reduction process
protein homotrimerization
cellular response to lipid hydroperoxide
glutathione transferase activity
glutathione peroxidase activity
transferase activity
identical protein binding
protein homodimerization activity
glutathione binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0