This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01261601_g1
See other ALDH7A1 GE Assays ›
Gene Symbol:
Aldh7a1
Gene Name:
aldehyde dehydrogenase family 7, member A1
Gene Aliases:
Atq1, D18Wsu181e
Chromosome Location:
Chr.18: 56524722 - 56572939 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01261601_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh7a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127338.1 NP_001120810.1 15 - 16 1468 94
NM_138600.4 NP_613066.2 13 - 14 1274 94
GenBank mRNA AK004991.1 - 15 - 16 1402 94
AK145873.1 - 13 - 14 1217 94
AK160117.1 - 13 - 14 1201 94
AK169195.1 - 13 - 14 1217 94
BC012407.1 - 13 - 14 1236 4191228 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldh7a1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
oxidation-reduction process
aldehyde dehydrogenase (NAD) activity
L-aminoadipate-semialdehyde dehydrogenase activity
betaine-aldehyde dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6s775:80/100.66.79.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0