This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01261866_g1
See other PPIL3 GE Assays ›
Gene Symbol:
Ppil3
Gene Name:
peptidylprolyl isomerase (cyclophilin)-like 3
Gene Aliases:
2310076N22Rik, 2510026K04Rik, Cyp10l
Chromosome Location:
Chr.1: 58430993 - 58445527 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01261866_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppil3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285826.1 NP_001272755.1 1 - 2 670 75
GenBank mRNA BC061645.1 - 1 - 2 613 5032617 75
DV043111.1 - 1 - 2 371 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC061645.1, NM_001285826.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
mRNA processing
protein folding
RNA splicing
peptidyl-prolyl cis-trans isomerase activity
isomerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100019382.mm.1 NM_001285826.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019382.mm.1 TC0100002382.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline