This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262200_m1
See other NDUFS1 GE Assays ›
Gene Symbol:
Ndufs1
Gene Name:
NADH dehydrogenase (ubiquinone) Fe-S protein 1
Gene Aliases:
5830412M15Rik, 9930026A05Rik
Chromosome Location:
Chr.1: 63143592 - 63176822 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262200_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufs1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160038.1 NP_001153510.1 3 - 4 570 102
NM_001160039.1 NP_001153511.1 3 - 4 412 102
NM_001160040.1 NP_001153512.1 2 - 3 466 102
NM_145518.2 NP_663493.2 2 - 3 308 102
GenBank mRNA AK034597.1 - 2 - 3 377 102
AK036926.1 - 2 - 3 173 102
AK078590.1 - 2 - 3 154 102
AK142711.1 - 2 - 3 188 102
AK146814.1 - 2 - 3 326 102
AK166604.1 - 3 - 4 259 102
AK167705.1 - 2 - 3 156 102
AK178076.1 - 1 - 2 78 102
BC006660.1 - 2 - 3 195 3501043 102
BC015300.1 - 2 - 3 128 4236882 102
BG808119.1 - 2 - 3 306 102
BU708393.1 - 3 - 4 413 102
BY709888.1 - 2 - 3 309 102
CN715066.1 - 2 - 3 184 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002445.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufs1

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

transport
apoptotic mitochondrial changes
ATP synthesis coupled electron transport
cellular respiration
ATP metabolic process
regulation of mitochondrial membrane potential
oxidation-reduction process
reactive oxygen species metabolic process
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity
electron carrier activity
oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H
metal ion binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100019908.mm.1 NM_001160038.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019908.mm.1 NM_001160038.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019908.mm.1 NM_001160039.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019908.mm.1 NM_001160039.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100019908.mm.1 TC0100002445.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0