This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262412_m1
See other NAPG GE Assays ›
Gene Symbol:
Napg
Gene Name:
N-ethylmaleimide sensitive fusion protein attachment protein gamma
Gene Aliases:
2400003O04Rik, SNARE
Chromosome Location:
Chr.18: 62977841 - 62999450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262412_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Napg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028017.1 NP_082293.1 8 - 9 557 73
GenBank mRNA AK010275.1 - 8 - 9 610 73
AK090379.1 - 8 - 9 519 73
AK154573.1 - 8 - 9 577 73
AK164149.1 - 8 - 9 552 73
AK167372.1 - 8 - 9 564 73
BC026977.1 - 8 - 9 557 5362684 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000648.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
vesicle-mediated transport
membrane fusion
soluble NSF attachment protein activity
syntaxin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800004786.mm.1 NM_028017.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004786.mm.1 NM_028017.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004786.mm.1 TC1800000648.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800004787.mm.1 NM_028017.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004787.mm.1 NM_028017.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004787.mm.1 TC1800000648.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0