This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262471_g1
See other MBD1 GE Assays ›
Gene Symbol:
Mbd1
Gene Name:
methyl-CpG binding domain protein 1
Gene Aliases:
Cxxc3, PCM1
Chromosome Location:
Chr.18: 74267615 - 74282734 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262471_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mbd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013594.2 NP_038622.2 12 - 13 1676 154
GenBank mRNA AF072240.1 - 14 - 15 1823 154
AK004624.2 - 12 - 13 1677 154
AK032392.1 - 4 - 5 620 154
AK032535.1 - 9 - 10 1174 154
AK156401.1 - 13 - 14 1667 154
AK166042.1 - 8 - 9 996 154
BC069837.1 - 12 - 13 1692 30435760 154

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mbd1

Gene Ontology Categories:

Function(s) Process(es)

DNA methylation
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of gene expression
negative regulation of astrocyte differentiation
DNA binding
chromatin binding
protein binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0