This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262511_m1
See other PSMG2 GE Assays ›
Gene Symbol:
Psmg2
Gene Name:
proteasome (prosome, macropain) assembly chaperone 2
Gene Aliases:
1700017I17Rik, AW545363, Clast3, Tnfsf5ip1
Chromosome Location:
Chr.18: 67641596 - 67654181 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262511_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Psmg2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134138.1 NP_598899.1 2 - 3 314 123
GenBank mRNA AB031387.1 - 2 - 3 314 123
AK077569.1 - 2 - 3 301 123
AK135578.1 - 2 - 3 306 123
AK167840.1 - 2 - 3 555 123
BC016606.1 - 2 - 3 293 2646038 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000708.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Psmg2

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

mitotic spindle assembly checkpoint
negative regulation of apoptotic process
proteasome assembly
regulation of cell cycle
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800005188.mm.1 NM_134138.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005188.mm.1 NM_134138.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800005188.mm.1 TC1800000708.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800005189.mm.1 TC1800000708.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t8jrk:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0