This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262532_m1
See other TCF4 GE Assays ›
Gene Symbol:
Tcf4
Gene Name:
transcription factor 4
Gene Aliases:
5730422P05Rik, ASP-I2, E2-2, E2.2, ITF-2, ITF-2b, ITF2, ME2, MITF-2A, MITF-2B, SEF-2, SEF2, SEF2-1, TFE, Tcf-4, bHLHb19
Chromosome Location:
Chr.18: 69344490 - 69687967 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262532_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083967.1 NP_001077436.1 8 - 9 785 82
NM_013685.2 NP_038713.1 8 - 9 1100 82
GenBank mRNA AK028748.1 - 8 - 9 711 82
AK081012.1 - 8 - 9 737 82
AK133885.1 - 9 - 10 1512 82
BC043050.1 - 7 - 8 560 5708370 82
S75870.1 - 8 - 9 672 82
U16322.1 - 8 - 9 1072 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf4

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
multicellular organism development
nervous system development
cell differentiation
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
chromatin binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
protein C-terminus binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
protein dimerization activity
E-box binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0