This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262554_g1
See other STARD6 GE Assays ›
Gene Symbol:
Stard6
Gene Name:
StAR-related lipid transfer (START) domain containing 6
Gene Aliases:
1700011K09Rik, 4833424I06Rik, 4933429L05Rik, AI644424
Chromosome Location:
Chr.18: 70472454 - 70505254 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262554_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stard6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289648.1 NP_001276577.1 4 - 5 376 77
NM_001289649.1 NP_001276578.1 4 - 5 355 77
NM_001289650.1 NP_001276579.1 4 - 5 390 77
NM_029019.4 NP_083295.1 4 - 5 445 77
GenBank mRNA AF480303.1 - 1 - 2 92 77
AK014759.1 - 4 - 5 356 77
AK016984.1 - 4 - 5 356 77
BC061022.1 - 2 - 3 128 6774873 77
BC087893.1 - 4 - 5 367 6774832 77
JF440953.1 - 2 - 3 164 77

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stard6

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
biological_process
lipid binding
cholesterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline