This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262605_m1
See other HSD17B4 GE Assays ›
Gene Symbol:
Hsd17b4
Gene Name:
hydroxysteroid (17-beta) dehydrogenase 4
Gene Aliases:
17-beta-HSD, 17[b]-HSD, 4, DBP, MFE-2, MFP2, MPF-2, Mfp-2, perMFE-2
Chromosome Location:
Chr.18: 50128201 - 50196270 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262605_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hsd17b4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008292.4 NP_032318.2 16 - 17 1586 67
GenBank mRNA AK004866.1 - 16 - 17 1586 67
AK088381.1 - 16 - 17 1561 67
AK147182.1 - 16 - 17 1558 67
AK152532.1 - 16 - 17 1480 67
AK161655.1 - 16 - 17 1481 67
AK166801.1 - 16 - 17 1499 67
AK167060.1 - 16 - 17 1536 67
AK169077.1 - 16 - 17 1488 67
AK215381.1 - 1 - 2 107 67
AK215718.1 - 1 - 2 107 67
BC022175.1 - 16 - 17 1459 4209394 67
X89998.1 - 16 - 17 1447 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000509.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hsd17b4

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
osteoblast differentiation
lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
metabolic process
androgen metabolic process
estrogen metabolic process
very long-chain fatty-acyl-CoA metabolic process
medium-chain fatty-acyl-CoA metabolic process
oxidation-reduction process
Sertoli cell development
3-hydroxyacyl-CoA dehydrogenase activity
receptor binding
oxidoreductase activity
long-chain-enoyl-CoA hydratase activity
lyase activity
isomerase activity
3alpha,7alpha,12alpha-trihydroxy-5beta-cholest-24-enoyl-CoA hydratase activity
protein homodimerization activity
17-beta-hydroxysteroid dehydrogenase (NAD+) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800003738.mm.1 NM_008292.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800003738.mm.1 NM_008292.4 Refseq
Affymetrix Mouse Clariom S Array PSR1800003738.mm.1 TC1800000509.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0