This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01262759_m1
See other NDST1 GE Assays ›
Gene Symbol:
Ndst1
Gene Name:
N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1
Gene Aliases:
1200015G06Rik, Hsst, NDST-1, b2b2230Clo
Chromosome Location:
Chr.18: 60684494 - 60748892 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01262759_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndst1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008306.4 NP_032332.2 8 - 9 2002 80
GenBank mRNA AF049894.1 - 8 - 9 1848 80
AF074926.1 - 8 - 9 1848 80
AK147282.1 - 9 - 10 2395 80
BC066098.1 - 8 - 9 2002 30533342 80
BC079561.1 - 8 - 9 2002 30533342 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001404.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndst1

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
polysaccharide biosynthetic process
protein deacetylation
protein sulfation
inflammatory response
smoothened signaling pathway
respiratory gaseous exchange
metabolic process
fibroblast growth factor receptor signaling pathway
embryo development
organ morphogenesis
glycosaminoglycan metabolic process
forebrain development
midbrain development
embryonic neurocranium morphogenesis
embryonic viscerocranium morphogenesis
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
catalytic activity
GPI-anchor transamidase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
[heparan sulfate]-glucosamine N-sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
hydrolase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
deacetylase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
coenzyme F390-A hydrolase activity
coenzyme F390-G hydrolase activity
hydroxyjasmonate sulfotransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0