This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01263578_m1
See other ELAVL4 GE Assays ›
Gene Symbol:
Elavl4
Gene Name:
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D)
Gene Aliases:
Elav, Hud, PNEM
Chromosome Location:
Chr.4: 110203737 - 110351946 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01263578_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elavl4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163399.1 NP_001156871.1 1 - 2 358 75
NM_010488.4 NP_034618.2 1 - 2 358 75
GenBank mRNA AK014133.1 - 1 - 2 126 75
BB648788.1 - 1 - 2 359 75
BC048159.1 - 1 - 2 331 5703005 75
D31953.1 - 1 - 2 105 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK014133.1, BC048159.1, D31953.1, NM_001163399.1, NM_010488.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Elavl4

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

learning
locomotory behavior
neuron differentiation
regulation of mRNA stability
dendrite morphogenesis
positive regulation of dendrite development
nucleotide binding
nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding
AU-rich element binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400026120.mm.1 TC0400003275.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026120.mm.1 NM_001163399.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026120.mm.1 NM_001163399.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026120.mm.1 NM_010488.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026120.mm.1 NM_010488.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400026121.mm.1 TC0400003275.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026121.mm.1 NM_001163399.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026121.mm.1 NM_001163399.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026121.mm.1 NM_010488.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026121.mm.1 NM_010488.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400026119.mm.1 TC0400003275.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026119.mm.1 NM_001163399.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026119.mm.1 NM_001163399.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026119.mm.1 NM_010488.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026119.mm.1 NM_010488.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 926125 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0