This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01263581_m1
See other ELAVL4 GE Assays ›
Gene Symbol:
Elavl4
Gene Name:
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D)
Gene Aliases:
Elav, Hud, PNEM
Chromosome Location:
Chr.4: 110203737 - 110351946 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01263581_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elavl4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163399.1 NP_001156871.1 6 - 7 1134 100
GenBank mRNA AK014133.1 - 6 - 7 902 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003275.mm.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

learning
locomotory behavior
neuron differentiation
regulation of mRNA stability
dendrite morphogenesis
positive regulation of dendrite development
nucleotide binding
nucleic acid binding
RNA binding
mRNA binding
mRNA 3'-UTR binding
AU-rich element binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400026100.mm.1 TC0400003275.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026100.mm.1 NM_001038698.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026100.mm.1 NM_001038698.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026100.mm.1 NM_001163397.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026100.mm.1 NM_001163397.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026100.mm.1 NM_001163399.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026100.mm.1 NM_001163399.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026100.mm.1 NM_010488.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026100.mm.1 NM_010488.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400026099.mm.1 TC0400003275.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026099.mm.1 NM_001038698.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026099.mm.1 NM_001038698.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026099.mm.1 NM_001163397.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026099.mm.1 NM_001163397.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026099.mm.1 NM_001163399.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026099.mm.1 NM_001163399.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026099.mm.1 NM_010488.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026099.mm.1 NM_010488.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400026101.mm.1 TC0400003275.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026101.mm.1 NM_001038698.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026101.mm.1 NM_001038698.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026101.mm.1 NM_001163397.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026101.mm.1 NM_001163397.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026101.mm.1 NM_001163399.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026101.mm.1 NM_001163399.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026101.mm.1 NM_010488.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026101.mm.1 NM_010488.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400026102.mm.1 TC0400003275.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026102.mm.1 NM_001038698.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026102.mm.1 NM_001038698.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026102.mm.1 NM_001163397.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026102.mm.1 NM_001163397.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026102.mm.1 NM_001163399.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026102.mm.1 NM_001163399.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026102.mm.1 NM_010488.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026102.mm.1 NM_010488.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400026103.mm.1 TC0400003275.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400026103.mm.1 NM_001038698.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026103.mm.1 NM_001038698.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026103.mm.1 NM_001163397.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026103.mm.1 NM_001163397.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026103.mm.1 NM_001163399.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026103.mm.1 NM_001163399.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026103.mm.1 NM_010488.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400026103.mm.1 NM_010488.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wt4p8:80/100.66.75.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0