This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01264040_m1
See other PCDH1 GE Assays ›
Gene Symbol:
Pcdh1
Gene Name:
protocadherin 1
Gene Aliases:
2010005A06Rik, AI585920
Chromosome Location:
Chr.18: 38185914 - 38212048 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01264040_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcdh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029357.3 NP_083633.2 1 - 2 202 79
GenBank mRNA AY861431.1 - 1 - 2 113 79
BC040402.1 - 1 - 2 202 4162978 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY861431.1, BC040402.1, NM_029357.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcdh1

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800009122.mm.1 NM_029357.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800009122.mm.1 NM_029357.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800009122.mm.1 TC1800001227.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800009123.mm.1 NM_029357.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800009123.mm.1 NM_029357.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800009123.mm.1 TC1800001227.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8mgvv:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0