This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01265378_g1
See other NPC1 GE Assays ›
Gene Symbol:
Npc1
Gene Name:
Niemann-Pick type C1
Gene Aliases:
A430089E03Rik, C85354, D18Ertd139e, D18Ertd723e, lcsd, nmf164, spm
Chromosome Location:
Chr.18: 12189694 - 12236386 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01265378_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Npc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008720.2 NP_032746.2 17 - 18 2752 79
GenBank mRNA AF003348.1 - 17 - 18 2729 79
AK154282.1 - 17 - 18 2672 79
AK155379.1 - 17 - 18 2672 79
BC052437.1 - 17 - 18 2752 6405214 79
BC054539.1 - 17 - 18 2752 6405214 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Npc1

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
lipid metabolic process
endocytosis
autophagy
adult walking behavior
steroid metabolic process
cholesterol metabolic process
negative regulation of macroautophagy
cholesterol transport
membrane raft organization
cholesterol efflux
response to drug
cholesterol homeostasis
response to cadmium ion
viral entry into host cell
negative regulation of cell death
cellular response to steroid hormone stimulus
cellular response to low-density lipoprotein particle stimulus
establishment of protein localization to membrane
positive regulation of cholesterol homeostasis
protein binding
hedgehog receptor activity
cholesterol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline