This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01265725_g1
See other POLR3C GE Assays ›
Gene Symbol:
Polr3c
Gene Name:
polymerase (RNA) III (DNA directed) polypeptide C
Gene Aliases:
4933407E01Rik, RPC3, RPC62
Chromosome Location:
Chr.3: 96711520 - 96727480 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01265725_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr3c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028925.1 NP_083201.1 6 - 7 887 76
GenBank mRNA AK016716.1 - 6 - 7 914 76
AK043194.1 - 5 - 6 1022 76
BC147485.1 - 6 - 7 887 9056238 76
BC151028.1 - 6 - 7 887 9087941 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002527.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

immune system process
transcription, DNA-templated
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA binding
DNA-directed RNA polymerase activity
transferase activity
nucleotidyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300019688.mm.1 NM_028925.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019688.mm.1 NM_028925.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019688.mm.1 TC0300002527.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300019685.mm.1 NM_028925.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019685.mm.1 NM_028925.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019685.mm.1 TC0300002527.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0