This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01265857_m1
See other NAPG GE Assays ›
Gene Symbol:
Napg
Gene Name:
N-ethylmaleimide sensitive fusion protein attachment protein gamma
Gene Aliases:
2400003O04Rik, SNARE
Chromosome Location:
Chr.18: 62977841 - 62999450 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01265857_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Napg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028017.1 NP_082293.1 5 - 6 309 91
GenBank mRNA AK010275.1 - 5 - 6 362 91
AK090379.1 - 5 - 6 271 91
AK154573.1 - 5 - 6 329 91
AK164149.1 - 5 - 6 304 91
AK167372.1 - 5 - 6 316 91
BC026977.1 - 5 - 6 309 5362684 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000648.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
vesicle-mediated transport
membrane fusion
soluble NSF attachment protein activity
syntaxin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800004780.mm.1 NM_028017.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004780.mm.1 NM_028017.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800004780.mm.1 TC1800000648.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0