This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01266028_m1
See other CBLL1 GE Assays ›
Gene Symbol:
Cbll1
Gene Name:
Casitas B-lineage lymphoma-like 1
Gene Aliases:
AI467391, Hakai, c-Cbl-like
Chromosome Location:
Chr.12: 31484829 - 31500741 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01266028_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cbll1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253847.1 NP_001240776.1 1 - 2 89 64
NM_001253848.1 NP_001240777.1 1 - 2 89 64
NM_134048.2 NP_598809.1 1 - 2 89 64
GenBank mRNA AF167441.1 - 1 - 2 89 64
AK049141.1 - 1 - 2 28 64
AK051737.1 - 1 - 2 28 64
AK166017.1 - 1 - 2 31 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF167441.1, AK049141.1, AK166017.1, NM_001253847.1, NM_001253848.1, NM_134048.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cbll1

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell adhesion
single organismal cell-cell adhesion
protein ubiquitination
positive regulation of cell migration
positive regulation of endocytosis
protein binding
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
identical protein binding
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
ubiquitin protein ligase activity
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200011055.mm.1 NM_001253847.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011055.mm.1 NM_001253847.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011055.mm.1 NM_001253848.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011055.mm.1 NM_001253848.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011055.mm.1 NM_134048.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011055.mm.1 NM_134048.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200011055.mm.1 TC1200001594.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200011057.mm.1 NM_001253847.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011057.mm.1 NM_001253847.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011057.mm.1 NM_001253848.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011057.mm.1 NM_001253848.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011057.mm.1 NM_134048.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200011057.mm.1 NM_134048.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200011057.mm.1 TC1200001594.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 370732 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0