This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01266218_m1
See other CPTP GE Assays ›
Gene Symbol:
Cptp
Gene Name:
ceramide-1-phosphate transfer protein
Gene Aliases:
Gltpd1
Chromosome Location:
Chr.4: 155864723 - 155869440 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01266218_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cptp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024472.4 NP_077792.2 1 - 2 274 109
GenBank mRNA AK040664.1 - 1 - 2 275 109
AK089229.1 - 1 - 2 244 109

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK040664.1, NM_024472.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cptp

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
glycolipid transport
ceramide 1-phosphate transport
phospholipid binding
phospholipid transporter activity
lipid binding
glycolipid transporter activity
glycolipid binding
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400034924.mm.1 TC0400004158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034924.mm.1 NM_024472.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034924.mm.1 NM_024472.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400034922.mm.1 TC0400004158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034922.mm.1 NM_024472.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034922.mm.1 NM_024472.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400034918.mm.1 TC0400004158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034918.mm.1 NM_024472.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034918.mm.1 NM_024472.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400034919.mm.1 TC0400004158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034919.mm.1 NM_024472.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034919.mm.1 NM_024472.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400034920.mm.1 TC0400004158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034920.mm.1 NM_024472.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034920.mm.1 NM_024472.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400034923.mm.1 TC0400004158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034923.mm.1 NM_024472.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034923.mm.1 NM_024472.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0