This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01266219_g1
See other CPTP GE Assays ›
Gene Symbol:
Cptp
Gene Name:
ceramide-1-phosphate transfer protein
Gene Aliases:
Gltpd1
Chromosome Location:
Chr.4: 155864723 - 155869440 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01266219_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cptp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024472.4 NP_077792.2 2 - 3 493 96
GenBank mRNA AK040664.1 - 2 - 3 494 96
BC002216.1 - 1 - 2 185 3490501 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400004158.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cptp

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

transport
lipid transport
glycolipid transport
ceramide 1-phosphate transport
phospholipid binding
phospholipid transporter activity
lipid binding
glycolipid transporter activity
glycolipid binding
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400034915.mm.1 TC0400004158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034915.mm.1 NM_024472.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034915.mm.1 NM_024472.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400034914.mm.1 TC0400004158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034914.mm.1 NM_024472.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034914.mm.1 NM_024472.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400034916.mm.1 TC0400004158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034916.mm.1 NM_024472.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034916.mm.1 NM_024472.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0