This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01266300_m1
See other URI1 GE Assays ›
Gene Symbol:
Uri1
Gene Name:
URI1, prefoldin-like chaperone
Gene Aliases:
C80913, NNX3, Rmp
Chromosome Location:
Chr.7: 37959992 - 38023575 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01266300_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Uri1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011274.5 NP_035404.4 2 - 3 473 85
GenBank mRNA AK133383.1 - 2 - 3 148 85
AK160558.1 - 2 - 3 413 85
AK166566.1 - 2 - 3 464 85
AK170929.1 - 2 - 3 468 85
BC023029.1 - 2 - 3 133 5374731 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK133383.1, AK170929.1, BC023029.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Uri1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of cell growth
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
response to virus
negative regulation of phosphatase activity
cellular response to growth factor stimulus
cellular response to steroid hormone stimulus
negative regulation of intrinsic apoptotic signaling pathway
RNA polymerase II transcription corepressor activity
chromatin binding
protein phosphatase inhibitor activity
phosphatase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700025696.mm.1 TC0700002873.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700025696.mm.1 NM_011274.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700025696.mm.1 NM_011274.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0