This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01266519_m1
See other PLCH2 GE Assays ›
Gene Symbol:
Plch2
Gene Name:
phospholipase C, eta 2
Gene Aliases:
A930027K05Rik, PLCeta2, Plc-eta2, Plcl4
Chromosome Location:
Chr.4: 154983115 - 155064091 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01266519_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plch2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113360.2 NP_001106831.1 13 - 14 2079 91
NM_175556.4 NP_780765.2 13 - 14 2079 91
GenBank mRNA AK129147.1 - 13 - 14 1839 91
AY966876.1 - 13 - 14 1857 91
BC040465.1 - 1 - 2 90 91
BC052329.1 - 3 - 4 212 91
DQ176851.1 - 13 - 14 2079 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400004133.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plch2

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
signal transduction
lipid catabolic process
intracellular signal transduction
phosphatidylinositol metabolic process
phosphatidylinositol phospholipase C activity
phospholipase C activity
signal transducer activity
calcium ion binding
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400034562.mm.1 TC0400004133.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400034562.mm.1 NM_001113360.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034562.mm.1 NM_001113360.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400034562.mm.1 NM_175556.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400034562.mm.1 NM_175556.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0