This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01267212_m1
See other PTPN2 GE Assays ›
Gene Symbol:
Ptpn2
Gene Name:
protein tyrosine phosphatase, non-receptor type 2
Gene Aliases:
AI325124, Ptpt, TC-PTP
Chromosome Location:
Chr.18: 67665501 - 67724748 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01267212_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127177.1 NP_001120649.1 1 - 2 178 103
NM_008977.3 NP_033003.1 1 - 2 178 103
GenBank mRNA AK014528.1 - 1 - 2 146 103
AK076072.1 - 1 - 2 131 103
AK132013.1 - 1 - 2 141 103
BC008269.1 - 1 - 2 152 3602983 103
M80739.1 - 1 - 2 133 103
M81477.1 - 1 - 2 133 103
S52655.1 - 1 - 2 128 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001512.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn2

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
negative regulation of cell proliferation
insulin receptor signaling pathway
negative regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of lipid storage
dephosphorylation
B cell differentiation
T cell differentiation
erythrocyte differentiation
peptidyl-tyrosine dephosphorylation
negative regulation of epidermal growth factor receptor signaling pathway
negative regulation of tyrosine phosphorylation of Stat3 protein
negative regulation of tyrosine phosphorylation of Stat5 protein
negative regulation of tyrosine phosphorylation of Stat6 protein
glucose homeostasis
negative regulation of macrophage differentiation
positive regulation of gluconeogenesis
negative regulation of insulin receptor signaling pathway
negative regulation of inflammatory response
negative regulation of T cell receptor signaling pathway
negative regulation of chemotaxis
negative regulation of interferon-gamma-mediated signaling pathway
negative regulation of type I interferon-mediated signaling pathway
negative regulation of protein tyrosine kinase activity
negative regulation of interleukin-6-mediated signaling pathway
negative regulation of ERK1 and ERK2 cascade
regulation of hepatocyte growth factor receptor signaling pathway
negative regulation of interleukin-2-mediated signaling pathway
negative regulation of prolactin signaling pathway
negative regulation of interleukin-4-mediated signaling pathway
negative regulation of macrophage colony-stimulating factor signaling pathway
negative regulation of positive thymic T cell selection
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of PERK-mediated unfolded protein response
negative regulation of platelet-derived growth factor receptor-beta signaling pathway
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
integrin binding
protein binding
hydrolase activity
phosphatase activity
protein kinase binding
syntaxin binding
receptor tyrosine kinase binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800011125.mm.1 NM_001127177.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011125.mm.1 NM_001127177.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011125.mm.1 NM_008977.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011125.mm.1 NM_008977.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800011125.mm.1 TC1800001512.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800011124.mm.1 NM_001127177.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011124.mm.1 NM_001127177.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011124.mm.1 NM_008977.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011124.mm.1 NM_008977.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800011124.mm.1 TC1800001512.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800011128.mm.1 NM_001127177.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011128.mm.1 NM_001127177.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011128.mm.1 NM_008977.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011128.mm.1 NM_008977.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800011128.mm.1 TC1800001512.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 421703 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0