This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01267215_m1
See other PTPN2 GE Assays ›
Gene Symbol:
Ptpn2
Gene Name:
protein tyrosine phosphatase, non-receptor type 2
Gene Aliases:
AI325124, Ptpt, TC-PTP
Chromosome Location:
Chr.18: 67665501 - 67724748 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01267215_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127177.1 NP_001120649.1 4 - 5 474 70
NM_008977.3 NP_033003.1 4 - 5 474 70
GenBank mRNA AK014528.1 - 4 - 5 442 70
AK076072.1 - 4 - 5 427 70
AK132013.1 - 4 - 5 437 70
BC008269.1 - 4 - 5 448 3602983 70
CA560046.1 - 4 - 5 474 70
M80739.1 - 4 - 5 429 70
M81477.1 - 4 - 5 429 70
S52655.1 - 4 - 5 424 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001512.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn2

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
negative regulation of cell proliferation
insulin receptor signaling pathway
negative regulation of tumor necrosis factor-mediated signaling pathway
negative regulation of lipid storage
dephosphorylation
B cell differentiation
T cell differentiation
erythrocyte differentiation
peptidyl-tyrosine dephosphorylation
negative regulation of epidermal growth factor receptor signaling pathway
negative regulation of tyrosine phosphorylation of Stat3 protein
negative regulation of tyrosine phosphorylation of Stat5 protein
negative regulation of tyrosine phosphorylation of Stat6 protein
glucose homeostasis
negative regulation of macrophage differentiation
positive regulation of gluconeogenesis
negative regulation of insulin receptor signaling pathway
negative regulation of inflammatory response
negative regulation of T cell receptor signaling pathway
negative regulation of chemotaxis
negative regulation of interferon-gamma-mediated signaling pathway
negative regulation of type I interferon-mediated signaling pathway
negative regulation of protein tyrosine kinase activity
negative regulation of interleukin-6-mediated signaling pathway
negative regulation of ERK1 and ERK2 cascade
regulation of hepatocyte growth factor receptor signaling pathway
negative regulation of interleukin-2-mediated signaling pathway
negative regulation of prolactin signaling pathway
negative regulation of interleukin-4-mediated signaling pathway
negative regulation of macrophage colony-stimulating factor signaling pathway
negative regulation of positive thymic T cell selection
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of PERK-mediated unfolded protein response
negative regulation of platelet-derived growth factor receptor-beta signaling pathway
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
non-membrane spanning protein tyrosine phosphatase activity
integrin binding
protein binding
hydrolase activity
phosphatase activity
protein kinase binding
syntaxin binding
receptor tyrosine kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800011119.mm.1 NM_001127177.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011119.mm.1 NM_001127177.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011119.mm.1 NM_008977.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011119.mm.1 NM_008977.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800011119.mm.1 TC1800001512.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0