This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01267917_m1
See other SLCO1C1 GE Assays ›
Gene Symbol:
Slco1c1
Gene Name:
solute carrier organic anion transporter family, member 1c1
Gene Aliases:
OATP-14, OATP-F, Oatp2, Oatpf, Slc21a14
Chromosome Location:
Chr.6: 141524378 - 141570177 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01267917_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco1c1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177772.1 NP_001171243.1 4 - 5 789 90
NM_021471.2 NP_067446.1 4 - 5 789 90
GenBank mRNA AK045782.1 - 792 90
AK048926.1 - 4 - 5 759 90
AK161010.1 - 759 90
AY007379.1 - 4 - 5 625 90
BC078456.1 - 4 - 5 739 30360663 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001671.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco1c1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
bile acid and bile salt transport
anion transmembrane transport
organic acid transmembrane transport
transporter activity
bile acid transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600013309.mm.1 TC0600001671.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600013309.mm.1 NM_001177772.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013309.mm.1 NM_001177772.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013309.mm.1 NM_021471.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013309.mm.1 NM_021471.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600013305.mm.1 TC0600001671.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600013305.mm.1 NM_001177772.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013305.mm.1 NM_001177772.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013305.mm.1 NM_021471.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013305.mm.1 NM_021471.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600013308.mm.1 TC0600001671.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600013308.mm.1 NM_001177772.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013308.mm.1 NM_001177772.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013308.mm.1 NM_021471.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013308.mm.1 NM_021471.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600013307.mm.1 TC0600001671.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600013307.mm.1 NM_001177772.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013307.mm.1 NM_001177772.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013307.mm.1 NM_021471.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013307.mm.1 NM_021471.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600013306.mm.1 TC0600001671.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600013306.mm.1 NM_001177772.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013306.mm.1 NM_001177772.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013306.mm.1 NM_021471.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600013306.mm.1 NM_021471.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0