This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01268258_g1
See other NAPA GE Assays ›
Gene Symbol:
Napa
Gene Name:
N-ethylmaleimide sensitive fusion protein attachment protein alpha
Gene Aliases:
1500039N14Rik, AW209189, RA81, SNAPA, SNARE, a-SNAP, hyh
Chromosome Location:
Chr.7: 16098456 - 16117975 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01268258_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Napa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025898.3 NP_080174.1 5 - 6 454 84
GenBank mRNA AK005372.1 - 5 - 6 456 84
AK013895.1 - 5 - 6 473 84
AK050623.1 - 5 - 6 518 84
AK136689.1 - 5 - 6 457 84
AK171998.1 - 5 - 6 518 84
BC004804.1 - 5 - 6 485 3584869 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000219.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Napa

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
brain development
regulation of synaptic vesicle priming
protein transport
vesicle-mediated transport
neuron differentiation
synaptic transmission, glutamatergic
SNARE complex disassembly
apical protein localization
membrane fusion
SNARE binding
soluble NSF attachment protein activity
protein binding
syntaxin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700001552.mm.1 TC0700000219.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700001552.mm.1 NM_025898.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001552.mm.1 NM_025898.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700001550.mm.1 TC0700000219.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700001550.mm.1 NM_025898.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700001550.mm.1 NM_025898.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l2rv2:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0