This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01268331_gH
See other SLC1A5 GE Assays ›
Gene Symbol:
Slc1a5
Gene Name:
solute carrier family 1 (neutral amino acid transporter), member 5
Gene Aliases:
AAAT, ASCT2, ATBO, M7V1, M7VS1, R16, RDRC, Slc1a7
Chromosome Location:
Chr.7: 16781345 - 16798274 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01268331_gH
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc1a5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009201.2 NP_033227.2 1 - 2 1132 65
GenBank mRNA AF246130.1 - 1 - 2 577 65
AK148346.1 - 1 - 2 1132 65
AK157838.1 - 1 - 2 1111 65
BC037462.1 - 1 - 2 723 4192790 65
L42115.1 - 1 - 2 1108 65

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc1a5

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
dicarboxylic acid transport
extracellular amino acid transport
neutral amino acid transport
L-serine transport
cation transmembrane transport
L-alpha-amino acid transmembrane transport
amino acid transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-serine transmembrane transporter activity
symporter activity
sodium:dicarboxylate symporter activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 919716 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0