This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01268553_m1
See other LIPG GE Assays ›
Gene Symbol:
Lipg
Gene Name:
lipase, endothelial
Gene Aliases:
3110013K01Rik, EL, lipase, mEDL
Chromosome Location:
Chr.18: 74939322 - 74961263 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01268553_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010720.3 NP_034850.3 1 - 2 384 84
GenBank mRNA AF118768.1 - 1 - 2 369 84
AK090224.1 - 1 - 2 382 84
AK152161.1 - 1 - 2 386 84
AK152430.1 - 1 - 2 382 84
AK152872.1 - 1 - 2 384 84
AK153541.1 - 1 - 2 386 84
AK172101.1 - 1 - 2 384 84
BC020991.1 - 1 - 2 238 3981856 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001595.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipg

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
cell proliferation
positive regulation of high-density lipoprotein particle clearance
lipid catabolic process
positive regulation of cholesterol transport
high-density lipoprotein particle remodeling
cholesterol homeostasis
reverse cholesterol transport
regulation of lipoprotein metabolic process
phospholipid homeostasis
lipoprotein lipase activity
phospholipase activity
triglyceride lipase activity
heparin binding
phosphatidylcholine 1-acylhydrolase activity
hydrolase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800011520.mm.1 NM_010720.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011520.mm.1 NM_010720.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800011520.mm.1 TC1800001595.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800011521.mm.1 NM_010720.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011521.mm.1 NM_010720.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800011521.mm.1 TC1800001595.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800011519.mm.1 NM_010720.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011519.mm.1 NM_010720.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800011519.mm.1 TC1800001595.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 371304 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0