This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01270320_m1
See other E2F6 GE Assays ›
Gene Symbol:
E2f6
Gene Name:
E2F transcription factor 6
Gene Aliases:
AI462434, E2F6b, EMA
Chromosome Location:
Chr.12: 16810903 - 16826752 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01270320_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: E2f6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033270.2 NP_150373.2 2 - 3 413 127
GenBank mRNA AB221552.1 - 2 - 3 174 127
AF032131.1 - 2 - 3 391 127
AK035116.1 - 2 - 3 410 127
BC057929.1 - 2 - 3 401 5289475 127
BY739130.1 - 2 - 3 414 127

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000132.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene E2f6

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription involved in G1/S transition of mitotic cell cycle
transcription, DNA-templated
regulation of transcription, DNA-templated
cell cycle
DNA binding
transcription factor activity, sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200001128.mm.1 NM_033270.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200001128.mm.1 NM_033270.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200001128.mm.1 NR_027677.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200001128.mm.1 NR_027677.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200001128.mm.1 TC1200000132.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0