This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01270678_m1
See other LDAH GE Assays ›
Gene Symbol:
Ldah
Gene Name:
lipid droplet associated hydrolase
Gene Aliases:
1110057K04Rik
Chromosome Location:
Chr.12: 8208100 - 8299566 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01270678_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ldah
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167767.1 NP_001161239.1 1 - 2 84 62
GenBank mRNA AI746795.1 - 1 - 2 84 62
AK036177.1 - 1 - 2 62 62
AK076460.1 - 1 - 2 75 62
AK162850.1 - 1 - 2 40 62
BC057311.1 - 1 - 2 64 6811681 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK076460.1, AK162850.1, BC057311.1, NM_001167767.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
lipid storage
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200000599.mm.1 NM_001167767.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000599.mm.1 NM_001167767.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000599.mm.1 NM_001167768.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000599.mm.1 NM_001167768.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000599.mm.1 NM_172401.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000599.mm.1 NM_172401.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200000599.mm.1 TC1200000058.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200000602.mm.1 NM_001167767.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000602.mm.1 NM_001167767.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000602.mm.1 NM_001167768.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000602.mm.1 NM_001167768.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000602.mm.1 NM_172401.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000602.mm.1 NM_172401.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200000602.mm.1 TC1200000058.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline