This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01270681_m1
See other LDAH GE Assays ›
Gene Symbol:
Ldah
Gene Name:
lipid droplet associated hydrolase
Gene Aliases:
1110057K04Rik
Chromosome Location:
Chr.12: 8208100 - 8299566 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01270681_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ldah
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172401.4 NP_765989.3 6 - 7 872 83
GenBank mRNA AK076460.1 - 4 - 5 538 83
AK079099.1 - 6 - 7 856 83
BC046986.1 - 5 - 6 791 6476504 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000058.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ldah

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
lipid storage
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200000611.mm.1 NM_001167767.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000611.mm.1 NM_001167767.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000611.mm.1 NM_172401.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200000611.mm.1 NM_172401.4 Refseq
Affymetrix Mouse Clariom S Array PSR1200000611.mm.1 TC1200000058.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 776722 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline