This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01270684_m1
See other SLC7A15 GE Assays ›
Gene Symbol:
Slc7a15
Gene Name:
solute carrier family 7 (cationic amino acid transporter, y+ system), member 15
Gene Aliases:
Arpat
Chromosome Location:
Chr.12: 8528481 - 8599066 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01270684_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc7a15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038660.2 NP_001033749.1 4 - 5 1176 72
NM_177802.3 NP_808470.1 5 - 6 733 72
GenBank mRNA AK033481.1 - 5 - 6 733 72
AY795939.1 - 4 - 5 1155 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001398.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc7a15

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
amino acid transport
L-alpha-amino acid transmembrane transport
protein binding
amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200009950.mm.1 NM_001038660.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200009950.mm.1 NM_177802.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200009950.mm.1 NM_177802.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200009950.mm.1 TC1200001398.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0