This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01270903_m1
See other PDIA6 GE Assays ›
Gene Symbol:
Pdia6
Gene Name:
protein disulfide isomerase associated 6
Gene Aliases:
1700015E05Rik, AL023058, C77895, CaBP5, P5, Txndc7
Chromosome Location:
Chr.12: 17266595 - 17284770 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01270903_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdia6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027959.3 NP_082235.1 1 - 2 55 78
GenBank mRNA AK005989.1 - 1 - 2 61 78
AK076558.1 - 1 - 2 56 78
AK133792.1 - 1 - 2 62 78
AK152960.1 - 1 - 2 68 78
AK166111.1 - 1 - 2 62 78
AK167386.1 - 1 - 2 73 78
AK168136.1 - 1 - 2 62 78
AK168279.1 - 1 - 2 62 78
BC006865.1 - 1 - 2 44 2645183 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK005989.1, AK076558.1, AK152960.1, AK166111.1, AK167386.1, AK168136.1, AK168279.1, BC006865.1, NM_027959.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdia6

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
response to endoplasmic reticulum stress
cell redox homeostasis
protein disulfide isomerase activity
isomerase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200001191.mm.1 NM_027959.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200001191.mm.1 NM_027959.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200001191.mm.1 TC1200000139.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200001190.mm.1 NM_027959.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200001190.mm.1 NM_027959.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200001190.mm.1 TC1200000139.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 703869 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline