This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01271413_g1
See other DCTPP1 GE Assays ›
Gene Symbol:
Dctpp1
Gene Name:
dCTP pyrophosphatase 1
Gene Aliases:
2410015N17Rik, AI854235, RS21-C6
Chromosome Location:
Chr.7: 127256959 - 127260667 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01271413_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dctpp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023203.1 NP_075692.1 2 - 3 252 74
GenBank mRNA AF110764.1 - 2 - 3 252 74
AK003643.1 - 2 - 3 231 74
AK010508.1 - 2 - 3 246 74
AK010606.1 - 2 - 3 219 74
BC004623.1 - 2 - 3 279 3585680 74

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dctpp1

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleoside triphosphate catabolic process
protein homotetramerization
nucleotide binding
magnesium ion binding
pyrophosphatase activity
hydrolase activity
pyrimidine deoxyribonucleotide binding
identical protein binding
metal ion binding
nucleoside-triphosphate diphosphatase activity
dCTP diphosphatase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504837 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504836 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 AK010606.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 BC004623.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 AK010508.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 NM_023203.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 AF110764.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 AK010606.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 BC004623.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 AK010508.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 NM_023203.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158876 AF110764.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39301 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309997 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76258 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108026 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 AK010606.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 BC004623.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 AK010508.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 NM_023203.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 AF110764.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 AK010606.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 BC004623.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 AK010508.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 NM_023203.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158876 AF110764.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504839 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504821 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504846 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504841 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504826 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504819 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504827 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504850 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504834 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504838 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504825 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504857 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504836 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504830 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504845 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504843 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504855 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504832 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504853 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504848 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504856 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504847 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504822 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504820 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25504829 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108026 - -
Whole Mouse Genome G4122A A_51_P158876 AK010606.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P158876 BC004623.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P158876 AK010508.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P158876 NM_023203.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P158876 AF110764.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P158876 AK010606.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P158876 BC004623.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P158876 AK010508.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P158876 NM_023203.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P158876 AF110764.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127712 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108026 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06490195 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06490191 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06490186 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06490190 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06490199 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06490197 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504836 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25504837 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504834 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504838 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504755 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504763 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504863 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504821 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504816 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504768 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504813 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504853 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504818 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504858 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504843 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504830 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504832 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504804 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504829 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504757 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504767 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504758 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504848 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504860 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504771 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504850 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504855 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25504808 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504868 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504847 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504830 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504841 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504834 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504832 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06133341 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504855 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06071382 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504810 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504808 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504813 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504803 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504839 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06392368 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06392370 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504858 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504821 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504869 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504804 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06314258 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504820 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06447705 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06392369 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504867 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05886445 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504846 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504850 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504818 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06068723 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06425383 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504845 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06133339 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504836 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504860 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504829 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504856 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504822 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06071383 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06412791 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504838 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504859 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504851 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504816 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504826 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25504861 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10568219 - -
Affymetrix Mouse Gene 1.0 ST Array 10568220 - -
Affymetrix Mouse Gene 1.0 ST Array 10568218 - -
Affymetrix HT MG-430 PM Array Plate 1416439_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10568217 - -
Mouse Genome 430 2.0 Array 1416439_at - -
Mouse Genome 430 2.0 Array 1416439_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66422_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66422_at - -
Affymetrix Mouse Clariom S Array PSR0700036286.mm.1 NM_023203.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036286.mm.1 NM_023203.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700036286.mm.1 TC0700004320.mm.1 MTA TC
Mouse Expression Set 430 1416439_at - -
Mouse Expression Set 430 1416439_PM_at - -
Murine Genome U74v2 Set 96293_at - -
Murine Genome U74v2 Set 96293_PM_at - -
Murine Genome U74v2 Set 165991_i_at - -
Murine Genome U74v2 Set 165991_PM_i_at - -
Mouse Genome 430A 2.0 Array 1416439_at - -
Mouse Genome 430A 2.0 Array 1416439_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1416439_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1416439_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 427106 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 4200707 - -
Illumina mouse-6 v1.0 expression beadchip 4200707 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004200707 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228334 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1228334 - -
mouse-6 v1.1 (Illumina) scl30652.3_263-S - -
Illumina Sentrix mouse-6.v1 bead array 4200707 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl30652.3_263-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200707 - -
MouseRef-8 v2 Expression BeadChip ILMN_1228334 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228334 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl30652.3_263-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0