This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273227_g1
See other PPP3CB GE Assays ›
Gene Symbol:
Ppp3cb
Gene Name:
protein phosphatase 3, catalytic subunit, beta isoform
Gene Aliases:
1110063J16Rik, Calnb, CnAbeta, Cnab
Chromosome Location:
Chr.14: 20499314 - 20546573 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273227_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp3cb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008914.2 NP_032940.1 2 - 3 420 108
GenBank mRNA AB221678.1 - 2 - 3 287 108
AK135203.1 - 2 - 3 234 108
BC037668.1 - 2 - 3 243 108
BC066000.1 - 2 - 3 391 5699179 108
M81483.1 - 2 - 3 257 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001540.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp3cb

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of T cell mediated cytotoxicity
lymphangiogenesis
protein phosphorylation
protein dephosphorylation
response to stress
heart development
regulation of gene expression
dephosphorylation
calcium ion regulated exocytosis
T cell differentiation
locomotion involved in locomotory behavior
calcineurin-NFAT signaling cascade
response to cytokine
cellular response to drug
positive regulation of insulin secretion involved in cellular response to glucose stimulus
T cell homeostasis
positive regulation of transcription from RNA polymerase II promoter
regulation of insulin secretion
positive regulation of NFAT protein import into nucleus
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
calcium-dependent protein serine/threonine phosphatase activity
calcium ion binding
protein binding
calmodulin binding
drug binding
hydrolase activity
enzyme binding
protein phosphatase 2B binding
calmodulin-dependent protein phosphatase activity
metal ion binding
protein heterodimerization activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400011532.mm.1 NM_008914.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400011532.mm.1 TC1400001540.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0