This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273564_m1
See other TFEB GE Assays ›
Gene Symbol:
Tfeb
Gene Name:
transcription factor EB
Gene Aliases:
Tcfeb, bHLHe35
Chromosome Location:
Chr.17: 47737037 - 47792416 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273564_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tfeb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161722.1 NP_001155194.1 1 - 2 282 55
GenBank mRNA AK078755.1 - 1 - 2 214 55

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK078755.1, NM_001161722.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus 1700 Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

embryonic placenta development
immune system process
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
autophagy
humoral immune response
lysosome organization
positive regulation of autophagy
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription regulatory region DNA binding
protein heterodimerization activity
protein dimerization activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 921685 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0