This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01273645_g1
See other PPP2R5D GE Assays ›
Gene Symbol:
Ppp2r5d
Gene Name:
protein phosphatase 2, regulatory subunit B', delta
Gene Aliases:
B'delta, TEG-271, Tex271
Chromosome Location:
Chr.17: 46682991 - 46705020 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01273645_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r5d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009358.3 NP_033384.2 13 - 14 1675 80
GenBank mRNA AB053464.1 - 13 - 14 1620 80
AF316616.2 - 13 - 14 1532 80
AK154050.1 - 13 - 14 1651 80
AK160587.1 - 13 - 14 1616 80
AY338234.1 - 13 - 14 1605 80
BC009163.1 - 3 - 4 202 80
BC010716.1 - 13 - 14 1588 2655861 80
X81059.1 - 2 - 3 117 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002186.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r5d

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
positive regulation of neuron projection development
regulation of protein autophosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of neurotrophin TRK receptor signaling pathway
cellular response to growth factor stimulus
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700020630.mm.1 NM_009358.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700020630.mm.1 NM_009358.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700020630.mm.1 TC1700002186.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline